Imprimer
Carboys, wide neck, LDPE, with screw cap and handles, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2234-0020 2234-0030 2234-0050
216-8721EA 91.5 EUR
216-8721 216-8722 216-8723
Carboys, wide neck, LDPE, with screw cap and handles, Nalgene®
Flacons Bonbonnes
Made of LDPE, translucent.
  • Leakproof white PP screw cap
  • Good chemical resistance
  • Good temperature resistance
Store and transport solids and powders with the wide mouth carboys with handles. A convenient, large opening promotes ease of cleaning and dispensing and can accommodate overhead mixers while its wide-shoulder handles enable easy carrying and pouring, even with gloved hands.