Imprimer
Control cell culture inserts, PET membrane, Corning® BioCoat™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
354572 354578
734-0215EA 352 EUR
734-0215 734-0221
Control cell culture inserts, PET membrane, Corning® BioCoat™
Microplaques Inserts pour plaques de cultures
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel® matrix, fibronectin, collagen and laminin.