Beryllium standard dissolved in oil, 1000 µg/g Be in mineral oil Specpure® for wear metal analysis
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Physical form: Liquid
MDL No.: MFCD00134032