Imprimer
Whatman™ Roby automated syringe filters
Fournisseur: Whatman products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10463806 10463898
514-0687EA 2690 EUR
514-0687 514-0695
Whatman™ Roby automated syringe filters
Filtres Filtres-seringue
Cytiva's Whatman Roby syringe filters are designed specifically for automated filtration systems. The design of the polypropylene housing ensures smooth transport from the storage turntable to the filtration site.
  • Optimized for SOTAX™, Caliper (Zymark™), Varian™, and Erweka™ tablet tester systems
  • Mechanically stable polypropylene housing specifically designed for automated filtration
  • Recyclable after use
Automated filtration using an autosampler greatly reduces the time and difficulty of sample preparation for tablet dissolution testing in a high-throughput laboratory.
Roby syringe filters have a polypropylene housing specifically designed for use in commonly available tablet tester systems to support this type of automated filtration.
Informations de livraison: le kit de validation de filtre Roby 25 inclut six types de filtres dans six tubes contenant chacun 25 filtres et le protocole de validation de filtre avec une aide pour le choix du filtre.