These portable microscopes are ideal for general purpose applications, where high image resolution and advanced hardware and software features are not strictly necessary.
- Economic, easy to use
- Magnification from 10 to 70× and around 200×
- Video frame rates up to 30 fps
- High speed USB 2.0 output
AM2111: Excellent choice for hobbyists and kids (use at schools).
AM3113T: The microscope features measurement functions and the Microtouch button to capture pictures directly from the Dino-Lite. The improved image sensor provides a high quality image with very natural colours and it also offers a high refresh rate for better real time images. It is an affordable solution for industrial, scientific and basic professional use.
Informations de commande: More models − UV/infrared illumination, higher magnification − upon request.
Informations de livraison: Supplied with software (DinoCapture 2.0 (Windows) DinoXcope (Mac OS)).