Imprimer
Handheld digital microscopes, Dino-Lite Basic
Fournisseur: Dino-Lite
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Dino-Lite® Basic
630-2934EA 145 EUR
630-2934 630-2675
Handheld digital microscopes, Dino-Lite Basic
Microscopes
These portable microscopes are ideal for general purpose applications, where high image resolution and advanced hardware and software features are not strictly necessary.
  • Economic, easy to use
  • Magnification from 10 to 70× and around 200×
  • Video frame rates up to 30 fps
  • High speed USB 2.0 output
AM2111: Excellent choice for hobbyists and kids (use at schools).
AM3113T: The microscope features measurement functions and the Microtouch button to capture pictures directly from the Dino-Lite. The improved image sensor provides a high quality image with very natural colours and it also offers a high refresh rate for better real time images. It is an affordable solution for industrial, scientific and basic professional use.
Informations de commande: More models − UV/infrared illumination, higher magnification − upon request.
Informations de livraison: Supplied with software (DinoCapture 2.0 (Windows) DinoXcope (Mac OS)).