Imprimer
Wash bottles, narrow or wide mouth, Standard Line
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
215-2496EA 40.6 EUR
215-2496 215-2506 215-2500 215-2498 215-2497 215-2534
Wash bottles, narrow or wide mouth, Standard Line
Bottles Wash Bottles
LDPE, translucent, with coloured, PP spray cap.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Colour coded caps for quick identification
Economical, soft and very easy to squeeze. For use with general solutions such as water, soap, dilute acids and alkalis. Dispensing tube with pointed end made of PP to optimise backflow of the medium.