Imprimer
VWR®, Combinaison de salle blanche Advanced SF
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
113-1068EA 173 EUR
113-1068 113-1073 113-1072 113-1071 113-1069 113-1067 113-1066 113-1065 113-1064 113-1063 113-1062 113-1061 113-1070
VWR®, Combinaison de salle blanche Advanced SF
Vêtements à usage unique
Les combinaisons VWR® Advanced SF sont fabriquées en textile de polypropylène spunbond, et revêtues d’un film de polyéthylène respirant.
  • Available with tapered collar or with hood and attached boot covers
  • Inset sleeves, zip closure, elasticated cuffs
  • Durable fabric, with good breathability and water transmission rate
  • Significant fluid and particulate barrier
  • Particle shedding: Level II Helmke drum classification
  • Soft, cloth-like fabric
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.