Imprimer
Papanicolaou solution (OG 6), Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
M-Clarity™ (Niveau de qualité) = MQ500
HT40116-500ML HT40132-1L
HT40116-500MLEA 52.4 EUR
HT40116-500ML HT40132-1L
Papanicolaou solution (OG 6), Sigma-Aldrich®
Papanicolaou solution (OG 6)
Orange G (certified), 0.3% and phosphotungstic acid, 0.015%, in denatured alcohol
Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.
Densité: 0,83 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00147833
UN: 1170
ADR: 3,II
IVDLogo