Imprimer
VWR®, Calibration Weights, Class 1
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
611-8759EA 86 EUR
611-8759 611-8760 611-8761 611-8762 611-8763 611-8764 611-8765 611-8766
VWR®, Calibration Weights, Class 1
Weights Calibration Weights
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
Informations de livraison: Individual weights are supplied without certificate.