Imprimer
Calibration weights, class E2
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
611-8391EA 398 EUR
611-8391 611-8392 611-8393 611-8394 611-8395 611-8396 611-8397 611-8101 611-8102 611-8103 611-8104 611-8105 611-8106 611-8107 611-8108 611-8109 611-8110 611-8111 611-8112 611-8113 611-8114 611-8115 611-8116 611-8117 611-8118 611-8119 611-8120 611-8121 611-8122 611-8398 611-8399 611-8400 611-8423 611-8424 611-8425 611-8426 611-8427 611-8131 611-8132 611-8133 611-8134 611-8135 611-8136 611-8137 611-8138 611-8139 611-8140 611-8141 611-8142 611-8143 611-8144 611-8145 611-8146 611-8147 611-8148 611-8149 611-8150 611-8151 611-8152 611-8428
Calibration weights, class E2
Poids Poids d'étalonnage
Weights made from solid, anti-magnetic, polished stainless steel. Suitable for high resolution electronic balances.
  • Wire weights up to 500 mg
  • Identification number guarantees traceability for weights from 1 g upwards
  • Available with or without COFRAC certificate
Conform to OILM R111 recommendations.
Certifications: Conform to OIML R111 recommendations.
Informations de commande: All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. A range of weight sets are also available - please search for product 'Calibration weight sets, Class E2'.
Cdt: Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.