Imprimer
Baths, thermostatically controlled, analogue, Clifton NE4-T series
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NE4-14T NE4-22T NE4-28T NE4-8T
462-0108EA 2750 EUR
462-0108 462-0109 462-0110 462-0107
Baths, thermostatically controlled, analogue, Clifton NE4-T series
Thermostats à circulation Systèmes à circulation ouverte Bains avec circulateurs détachables
These thermostatically controlled analogue stirred baths have a corrosion resistant stainless steel tank and durable outer case. Immersed components are corrosion resistant.
  • Thermostatic control, easy to use dial setting, ambient 5 °C to 99 °C
  • Powerful stirring within the tank for good water circulation and temperature stability
  • Controls with an illuminated on/off switch and “heater on” indicator
  • User resettable safety cut-out float switch for low liquid level protection
  • Anti-bacterial paint finish that actively inhibits bacterial growth