Imprimer
Accessoires et pièces de rechange
Fournisseur: VACUUBRAND
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20696810 20696811 20696812 20696813 20696814 20696815 20696819 20696821 20696840 20696841 20699926 20637777
181-1671EA 58.1 EUR
181-1671 181-1672 181-1673 181-1675 181-1676 181-1677 181-1674 181-1678 181-1621 181-1622 181-1653 VACU20637777
Accessoires et pièces de rechange
Pompe Pompes à vide à membrane
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.