Imprimer
Thermometer test caps
Fournisseur: COMARK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TX23L TX24L TX25L TX26L
620-0377EA 176 EUR
620-0377 620-0378 620-0379 620-0380
Thermometer test caps
Thermomètres Thermomètres numériques
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.