Imprimer
Hand-Drill, QualiRod
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5311-0001 5311-0002
300-0018EA 114 EUR
300-0018 300-0140
Hand-Drill, QualiRod
Samplers Solid Samplers
Conical hand-drill, conically shaped, with sharp cutting edges and a solid handle. Ideal to cut out samples from soft, paste-like substances (cheese, butter, clay, loam, soap, wax, etc.).
  • Stainless steel V2A (1.4301)/ AISI 304
  • Sterilisable
  • Conforms to standards EN/ISO 707
Total length 175 mm, weight 96 g.
Certifications: ISO 707