Imprimer
Sampler, StickProof
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5338-0410 5338-1100
312-1176EA 683 EUR
312-1176 300-0117
Sampler, StickProof
Samplers Solid Samplers
Stainless steel sampler for powders. The conical inlet allows for collection of different sample sizes, up to approximately 50 ml. Constructed without edges, grooves or crevices for easy cleaning. The sample is collected directly into the sampling bag which is held in place with the included clamp.
  • Especially slender tip can be inserted easily and deeply
  • Electropolished surface
  • Variable sample size up to 50 ml
  • Stainless steel V4A (1.4404)/AISI 316 L
Informations de livraison: Complete with clamp and 100 sampling bags PE.