Imprimer
Réfractomètres portables, série MASTER
Fournisseur: Atago
Ratings:
Moyenne des Evaluations: 5.0 (1 Avis)
Total Evaluation: 1
Moyenne des Notes : 5.0 sur 5
Cet avis vous a-t-il aidé ? Oui Non
Oui : 0 Non : 0 
chimie / laboratoire / eyrein
Oui, je le conseille
Appareil fiable et robuste-utilisation simple et rapide. attention à la fragilité du capot , pièce qui existe en accessoire.
2364 2374 2384 2363
635-0693EA 305 EUR
635-0693 635-0697 635-0701 635-0692
Réfractomètres portables, série MASTER
Réfractomètres Réfractomètres portables
The MASTER series of handheld refractometers provides a range of easy to use instruments for a wide variety of applications.
  • MASTER-M: Standard model, clear scale, only two drops required
  • MASTER-T: Automated temperature compensation from 10 to +30 °C
  • MASTER-α: Automated temperature compensation (IP 65 except eyepiece), dust-tight and protected against water jets
  • MASTER-P: Designed with a plastic body for resistance to salty and acidic samples
The design has an ergonomic grip and a prism housing that allows for the sample to cover the prism without having to lift the daylight plate.
These instruments have automated temperature compensation in the range 10 to 30 °C.