Imprimer
Repetitive pipette, Multipette® M4
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4982000.012 4982000.314
613-2890EA 461 EUR
613-2890 613-2927
Repetitive pipette, Multipette® M4
Pipettes Repetitive Pipettors
Manual dispenser, designed for maximum ease of use, precision and flexibility in terms of volume selection and replacement of the associated Combitip® Advanced tips. Combitip® Advanced tips work on the direct displacement principle, ensuring that the correct volume is dispensed every time, regardless of the density of the liquid or its flow properties. This principle also ensures safer conditions when working with radioactive, aggressive, toxic substances or liquids with higher vapour pressures.
  • Simple volume adjustment from 1 µl to 10 ml
  • Volume display with automatic recognition of the Combitip® Advanced tip
  • Up to 100 dispensing cycles with one filling, integrated step counter
  • Capable of dispensing 112 different volumes
  • The Combitip® can be detached by applying slight pressure, avoiding any contact with and contamination of the tip
Combitip advanced® tips work on the direct displacement principle, ensuring that the correct volume is dispensed every time, regardless of the density of the liquid or its flow properties. This principle also ensures safer conditions when working with radioactive, aggressive, toxic substances or liquids with higher vapour pressures.