Évolutifs, les SZX16 sont totalement dédiés à la recherche moderne. Ils bénéficient d’une résolution de jusqu’à 900 lignes par millimètre pour des observations encore plus précises. L'épifluorescence pour la recherche in vivo assure une luminosité optimale.
- Trajectoire d’illumination par fluorescence perpendiculaire, ce qui empêche l’apparition d’artéfacts
- Grand facteur de zoom et bonne perception du relief ce qui améliore la profondeur des observations
- Plage de grossissement comprise entre 3,5 et 230× sans interruption de l’observation
Optics, which include observation tubes, zoom body, and objectives are fully apochromatic to reduce chromatic blur over the entire magnification range. Objectives can deliver clear and high-contrast images of virtually any specimen. The maximum NA of 0,3 produces a resolution of 900 linepairs per millimeter allowing clear visualization of fine microscopic cell and cell component structures. Such amazing resolution and peerless magnification makes work more efficient, more precise and reveals much more information from samples.
With a slim 41,5 mm design, that is approximately half the thickness of previous halogen lamp transmitted light illumination bases, new LED transmitted light illumination bases have a lower height to enable a low eyepoint and easy access to base-mounted samples during observation and operation. The LED illumination base with quad position turret enables the user to choose cartridges and to switch from brightfield (standard/high/low), oblique (standard/high/low), darkfield, polarised illumination, and shutter with a simple turn. A one position LED illumination base is also an option. This makes the SZX16 a flexible all-in-one microscope for various samples and observation tasks. Another advantage of LED illumination is a cooler base surface, which is suitable for long duration manipulation of live specimens. Power consumption is lower than a conventional 30 W halogen light source. A life cycle of over 60000 hours significantly reduces operation costs.