Imprimer
UV inspection lamps, Compact series
Fournisseur: UVP ULTRA VIOLET PRODUCTS
Ratings:
Moyenne des Evaluations: 5.0 (1 Avis)
Total Evaluation: 1
Moyenne des Notes : 5.0 sur 5
Cet avis vous a-t-il aidé ? Oui Non
Oui : 0 Non : 0 
Academic/Research / Lecturer/Researcher / Lagos, Nigeria
Oui, je le conseille
A very useful device for my fractionation and isolation of medicinal plant. It was very resourceful in collection of eluate from column and spot identification on TLC plate. Hope I could get this in Lagos, Nigeria especially the dual 254/365nm 230V, as the one I used while in Chicago for research work was 115V.
Réponse
de VWR France Publié 01/02/2017
Thank you for taking the time to provide feedback on VWR.com. Your local VWR distributor, LS Scientific, will be able to help you with any purchasing needs. olumurewa.odunjo@lsscientific.com
95-0021-10 95-0016-15 95-0018-03 95-0017-10 95-0019-03
732-0859EA 494 EUR
732-0859 733-2366 730-1525 732-0858 730-1526
UV inspection lamps, Compact series
Lights UV Lights, Lamps and Bulbs
Made of durable, lightweight plastic, these compact 4 W UV lamps can be used as a handheld instrument or hands-free in combination with an optional stand. Suitable for a wide range of applications, including fluorescence of stained gels and sterilisation. UV tubes and filters can be replaced simply and conveniently.
  • Lightweight, easy to handle lamp design
  • On/off button conveniently located on the back of the lamp
  • Models available include 254 nm shortwave, 365 nm longwave or a combination shortwave/longwave (365/254 nm)
  • Shortwave/longwave version (UVGL-15) comes with a snap-on wavelength selector for switching between wavelengths
  • UVGL-25 with 1×254 nm and 1×365 nm UV tube can be switched from short to longwave
W×D×H: 198×71×53 mm
W×D×H: 198×71×53 mm
Weight: 0,45 kg