Imprimer
Disposable pestles, microtubes and cordless motor for pellet mixing
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
431-0094EA 267 EUR
431-0094 431-0095 431-0096 431-0098 431-0099
Disposable pestles, microtubes and cordless motor for pellet mixing
Homogenizers Handheld Homogenizers
Pestles and microtubes manufactured from polybutylene terephthalate (PBTP) in a class 10000, ISO Class 7, M5.5 cleanroom. Designed for homogenising cells and plant tissue or for re-suspending protein and DNA pellets. An optional battery powered motor can be used with the pestles and microtubes. Pestles are 75 mm in length.
  • Free from DNase, RNase and non pyrogenic
  • Grooved grip makes pestle handle easy to turn without slipping, even when wearing gloves
  • Lightweight motor reduces hand fatigue
  • High speed vortexing action of the motor completely homogenises cell tissue or pellets within seconds
Informations de livraison: Pestles and microtubes are individually wrapped. Optional motor is supplied with a pestle adapter and two AA batteries.