Imprimer
CentriVap® mobile systems
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7812011 7812014
531-0169EA 18960 EUR
531-0169 531-0170
CentriVap® mobile systems
Evaporators Centrifugal Evaporators
Combine the separate concentrator and cold trap components into one space-saving console and include casters for portability from laboratory to laboratory. A 12 to 17 mm rotor and solvent trap are also included. The interior can accommodate a vacuum pump - the only additional component necessary to begin processing.
  • Epoxy-coated aluminum chamber with glass lid
  • Microprocessor-controlled centrifugation up to 1725 rpm and microprocessor-controlled heating of 300 watts
  • Control panel with on / off button, preheat button, optional Heat Boost ™ button, parameter selection, three Quick-Start ™ buttons and activation of the cooling trap; Quick-Stop ™ system for immediate stopping
  • Outlet valve for automatic emptying
  • Large LCD screen for display of program number, current and set temperature, heating duration, cycle duration, activation of the optional Heat Boost ™ and selection of the cooling trap
Informations de livraison: Vacuum pump not included.