Imprimer
Dissolved oxygen probes, CellOx® / DurOx®
Fournisseur: WTW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
201533 201545 201546 201570
715-2917EA 1240 EUR
715-2917 664-0511 662-0391 664-0512
Dissolved oxygen probes, CellOx® / DurOx®
Électrodes Sondes à oxygène dissous
Galvanic dissolved oxygen sensors for universal (CellOx®) or field use (DurOx®).
  • Membrane-covered sensors
  • User friendly and long operation time
CellOx® 325 is a dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring, which can be used in the lab as well as in the field. Its features include a quick response time (t99<60 s) and low-maintenance operation. Its fields of use include surface waters, measurements in wastewater treatment plants, BOD directions, but also industrial and other applications. It is filled with electrolyte and achieves a long dwelling time up to 6 months. The calibration vessel is part of the delivery.
DurOx® is a robust, membrane-covered galvanic dissolved oxygen sensor for applications in flowing waters, fish hatcheries, aeration basins and more fields. With screw-on protection. The special membranes achieve stable measured values, especially in the aeration basin and similar applications with strongly fluctuating oxygen content.
Measuring ranges:
Oxygen: 0 to 50 mg/L (concentration); 0 to 600% (saturation)
Partial pressure: 0 to 1250 mbar