Imprimer
Conductivity cells, four-electrode, Orion™ DuraProbe™
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
013005MD 013010MD 013610MD 013605MD
663-0180EA 588 EUR
663-0180 663-0218 663-0220 663-0189
Conductivity cells, four-electrode, Orion™ DuraProbe™
Électrodes Électrodes de conductivité
These durable and extremely accurate conductivity cells can be used under a wide range of conditions. Intended for field or laboratory use.
  • Virtually eliminate fringe field interference errors
  • Rugged epoxy body
  • Graphite sensors
  • Temperature compensated