Imprimer
Arsenic determination apparatus, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AD3 1AD/1
BIBBAD3EA 237 EUR
BIBBAD3 201-8003
Arsenic determination apparatus, Quickfit®
Systèmes de digestion Appareil de détection de traces d'éléments
PYREX®, borosilicate glass arsenic determination apparatus designed for the estimation of arsenic and antimony.
  • Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 5380A
  • Complete assembly (all parts single available)