Imprimer
Chairs, Stools and stand-up seat, stainless steel
Fournisseur: Kango
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7NN12VHXX00905 7NN12VHXX04905 7KA82FXYO00512 7KA82FXYO00905 7NG26VHXX04905 7NG36VHXX04905 7NG36VHXX00905 7NG26VHXX00905
132-8451EA 764 EUR
132-8451 132-8452 139-0468 139-0207 BIEN7NG26VHXX04905 139-0440 BIEN7NG36VHXX00905 BIEN7NG26VHXX00905
Chairs, Stools and stand-up seat, stainless steel
Chairs
Chairs, stools and stand-up with polyurethane foam seats and backrests with stainless steel 316 L base.
  • 316L stainless steel column and base (five-branch base for the chairs and stools / horse shoe base with anti-slip coating for the stand-up seat)
  • Easily cleaned using common household products
Chairs
Ergonomically designed seats and backrests made from 40 mm thick, soft, comfortable polyurethane foam
No irritating sweating
Resistant to nicks, weak acids, sparks and lubricants
The 316L stainless steel base is especially corrosion-resistant
Height adjustment with screw hidden inside the column
Available with or without footrest
Fire resistant
Dimensions of the seat (W×D): 410×390 mm
Dimensions of the backrest (W×H): 360×220 mm
Stools
Round seat made from 40 mm thick, soft, comfortable polyurethane foam
Resistant to nicks, weak acids, sparks and lubricants
The 316L stainless steel base is especially corrosion-resistant
Ideal for use in wet rooms, cleanrooms, laboratories and in the food industry
Height adjustment with screw hidden inside the column
Available with or without footrest
Fire resistant
Diameter of the seat: 320 mm
Stand-up seat
Semicircular 316L stainless steel base with anti-slip PVC coating
Cylindrical seat in PU
Carrying handle built into the seat
Space-saving
With telescopic height adjustment
Dimensions of the seat (W×D): 345×100 mm