Imprimer
Semi-dry electroblotters, Owl™ HEP-1 and HEP-3
Fournisseur: THERMO OWL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HEP-1 HEP-3
730-0110EA 2360 EUR
730-0110 730-0111
Semi-dry electroblotters, Owl™ HEP-1 and HEP-3
Blotting Apparatus
These sleek, low profile systems are designed for efficient transfer of proteins and nucleic acids to membranes. The base contains a stainless steel cathode plate and the lid houses a platinum/titanium anode plate.
  • Requires minimal amount of buffer - the buffer contained within the blotting filter paper is sufficient for efficient transfer of molecules
  • Two models available for transfer of up to 4 or 8 mini gels, or a variety of larger gels
  • Transfers can be completed in as little as 15 minutes
  • Ideal for proteins under 150 kd