Imprimer
Sondes pH/ATC 3 en 1, Orion™ Triode™
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9107BN 9107BNMD 9107WMMD
662-0781EA 247 EUR
662-0781 662-1145 662-1453
Sondes pH/ATC 3 en 1, Orion™ Triode™
Électrodes Électrodes pH
These versatile 3-in-1 pH/ATC (automatic temperature compensation) probes can be used in either the field or a plant.
  • Low-maintenance, gel-filled electrode with built-in ATC
  • Non refillable
  • Epoxy body for ruggedness
  • Ag/AgCl internal reference system, sealed reference
pH range: 0 to 14
Recommended for use with:
9107BN & 9107WP: for Orion A/A+ models 210, 230, 250, 410, 420, 520, 525, 710, 720, & 920
9107BNMD, 9107WMMD, 9107WLMD: for Orion STAR A111, STAR A121, STAR A211, STAR A221, STAR A321, Dual Star, & Versa Star