Imprimer
Lingettes présaturées, PolySat®
Fournisseur: Texwipe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TX1051 TX1051_U
115-0301EA 799 EUR
115-0301 115-2829
Lingettes présaturées, PolySat®
Lingettes Lingettes et essuie-mains à usage unique Lingettes présaturées
PolySat® is constructed from a 100% melt-blown polypropylene substrate that is pre-wetted with 70% isopropyl alcohol and packaged in a resealable FlexPack. PolySat® is convenient for cleaning sensitive surfaces and polishing equipment in controlled environments.
  • FlexPacks are a convenient alternative to wipes and spray bottles, eliminating mixing, sterilising and waste
  • Packed in SolventSafe plastic that can be wiped down prior to introduction into the cleanroom
  • Delivers a pre-measured amount of high purity alcohol to the surface: Optimises alcohol usage and reduces spill potential
  • Low particle generation and low levels of solvent extractables
Couleur: blanc
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904