Imprimer
Watch glasses
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N634952430790 N634952430860 N634952430910 N634952430950 N634952430010 N634952430080
216-1172EA 2.7 EUR
216-1172 216-1175 216-1176 216-1177 216-1178 216-1180
Watch glasses
Watch Glasses
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fused edges.
Certifications: DIN 12341