Imprimer
Viscosimètres Ubbelohde (DIN)
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285400078 285400148 285400156 285400004 285400029 285400037 285400045 285400053 285400061 285400086 285400094 285400107 285400115 285400123 285400131
616-1108EA 617 EUR
616-1108 645-0069 616-0030 616-1101 616-1103 616-1104 616-1105 616-1106 616-1107 616-1109 616-1110 616-1111 616-1112 616-1113 616-0029
Viscosimètres Ubbelohde (DIN)
Viscosimètres Rhéomètres capillaires
Ubbelohde viscometers for the determination of absolute and relative viscosity of liquids with Newtonian flow behaviour.
  • Filling quantity: 15 - 20 ml
  • Recommended minimum flow through time is 200 seconds
  • Overall length ∼290 mm
Calibrated with constant for manual measurement.
Certifications: DIN 51562-1, ISO 3105.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable