Imprimer
Cryogenic vials
Fournisseur: VWR Collection
Ratings:
Moyenne des Evaluations: 5.0 (1 Avis)
Total Evaluation: 1
Moyenne des Notes : 5.0 sur 5
Cet avis vous a-t-il aidé ? Oui Non
Oui : 3 Non : 1 
Medical laboratory
Oui, je le conseille
We've been using those cryogrnic vials in our laboratory, they are excellent!
Réponse
de VWR France Publié 03/04/2025
We appreciate your feedback. Thank you.
479-1238EA 538 EUR
479-1238
Cryogenic vials
Vials
Designed for the storage of biological material, human or animal cells, at temperatures as low as –196 °C (should only be used in the gas phase of liquid nitrogen).
  • Graduations and white marking area
  • Sterilised by gamma radiation
  • Compatible with most storage systems
Closures and tubes are made of PP, and both have the same coefficient of expansion, further enhancing the leakproof qualities of these vials at various temperatures. Round bottom vials can be centrifuged up to 17000×g.
Round bottom vials can be centrifuged up to 17 000×g
Certifications: Certified RNase-, DNase-, pyrogen- and DNA-free.
Cdt: Packed in unique tamper-proof, resealable, safety-lock bags of 100.