Imprimer
Joints rodés sphériques
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
01201305 01201909 01202915 01203520 01204125 01211305 01211909 01212915 01213520 01214125
201-4212EA 12.3 EUR
201-4212 201-4213 201-4214 201-4215 201-0127 201-4222 201-4223 201-4224 201-0128 201-0129
Joints rodés sphériques
Adaptateurs Adaptateurs rodés sphériques
DURAN®, borosilicate glass, clear.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
Spherical cup and ball joint with inch markings. Fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving relatively high resistance to temperature changes.
DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641
Certifications: DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641.