Imprimer
HelixMark® Standard Silicone Tubing
Fournisseur: Freudenberg Medical Europe GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60-411-57 60-411-40 60-411-42 60-411-44 60-411-58 60-411-51 60-411-52 60-411-53 60-411-54 60-411-55 60-411-59 60-411-60 60-411-63 60-411-43 60-411-49 60-411-50 60-411-56 60-411-62 60-411-65 60-411-61B 60-411-46 60-411-47 60-411-41 60-411-45 60-411-48 60-411-64 60-411-61
228-0662EA 415 EUR
228-0662 228-0661 228-1152 228-1153 228-0663 228-1094 228-1095 228-1096 228-1097 228-1098 228-1100 228-1101 228-1104 228-1091 228-1092 228-1093 228-1099 228-0664 228-0845 228-1385 228-1382 228-1383 HELI60-411-41 HELI60-411-45 HELI60-411-48 HELI60-411-64 HELI60-411-61
HelixMark® Standard Silicone Tubing
Tuyaux Transfer Tubing
HelixMark standard silicone tubing meets the needs of a broad spectrum of medical and laboratory applications and are universally applicable for the transfer of fluids under standard requirements. The tubing is individually packed in 15 m coils.
  • Utilisation of platinum-cured silicone
  • Production and packaging in ISO Class 8 clean rooms
  • Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
  • Seamless batch traceability
  • Double packed
  • Customer-specific solutions available outside the HelixMark standard product portfolio
All HelixMark products are manufactured in clean rooms in compliance with the requirements of the medtech and pharmaceutical industry.
Typical hardness: 60 Shore
Density: 1.15
Certifications: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177.2600 and EP 3.1.9. Certified manufacturing in compliance with ISO 13485 quality standards. Supplied with material certificate in every packaging unit.