Imprimer
HelixMark® Braided Reinforced Tubing
Fournisseur: Freudenberg Medical Europe GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60-664-50 60-664-70 60-664-60 60-664-100 60-664-20 60-664-30 60-664-40 60-664-45 60-664-90 60-664-80 60-664-60A 60-664-60C
228-0846EA 1030 EUR
228-0846 228-0847 228-0665 228-1105 228-1106 228-1107 228-1108 228-1109 228-1110 228-0848 228-1386 228-1387
HelixMark® Braided Reinforced Tubing
Tuyaux Transfer Tubing
HelixMark braided reinforced tubing is designed and manufactured especially for applications with higher pressures. Pressure-proof polyester braiding, firmly embedded between two layers of silicone, lends this tubing its particularly durability and resistance to pressure. The tubing is individually packed in 7,5 m coils.
  • Utilisation of platinum-cured silicone
  • Production and packaging in ISO Class 8 clean rooms
  • Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
  • Seamless batch traceability
  • Double packed
  • Customer-specific solutions
En savoir plus