Imprimer
Tuyaux Tygon® S3™ E- LFL
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AVX42003 AVX42007 AVX42012 AVX42022 AVX42017 AVX42019 AVX42029 AVX42038 AVX06057 070602-08-ND 070602-09-ND 070602-10-ND 070602-13-ND 070602-06-ND 070602-18-ND 070603-05-ND 070603-12-ND
228-1258EA 530 EUR
228-1258 228-1259 228-1260 228-1262 228-1261 228-1321 228-1322 228-1323 228-1319 228-1329 228-1330 228-1331 228-1334 228-1327 228-1339 228-1343 228-1350
Tuyaux Tygon® S3™ E- LFL
Tuyaux Transfer Tubing
Tubes sans DEHP, ni phtalates
  • Over 1000 hours pump tubing life at 0 psi; over 600 hours pump tubing life at 10 psi
  • Extremely low particle spallation
  • Contains no BPA or phthalates
  • Broad chemical resistance
  • Meets USP Class VI, ISO 10993 and FDA criteria - suitable for pharma, medical and food applications
Longest flexible life of any clear Tygon® tubing. The non ageing and broad chemical resistance properties enable users to utilise the tubing for transferring chemicals, cleaning agents and bactericides without danger of corrosion (as with metals) or oxidation (as with rubber). Tygon® S3™ E-LFL tubing is formulated specifically for use in peristaltic pump applications.
Temperature resistant from −55 to +74 °C.
Hardness (Shore durometer A): 56.
Density: 1,17.
Length 381 mm, distance between stops 152 mm.