Imprimer
Gas inlet/air bleed tubes
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05083001 05083002 05083003 05083004
201-2621EA 24 EUR
201-2621 201-1069 201-1070 201-2622
Gas inlet/air bleed tubes
Tubes Gas Inlet Tubes
Duran® borosilicate 3.3 clear glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90° with fused cone joint for cutting or drawing off capillaries to a required length.
Total length: 250 mm