Imprimer
Round bottomed test tubes, Falcon®
Fournisseur: Corning
Ratings:
Moyenne des Evaluations: 5.0 (1 Avis)
Total Evaluation: 1
Moyenne des Notes : 5.0 sur 5
Cet avis vous a-t-il aidé ? Oui Non
Oui : 1 Non : 0 
CHIMIE / LABORATOIRE / EYREIN
Oui, je le conseille
ARTICLE QUI EST PRATIQUE ET SOLIDE ET COMPETITIF EN PRIX
352001 352003 352008 352017 352025 352027 352037 352045 352051 352052 352054 352057 352058 352235
734-0435EA 211 EUR
734-0435 734-0436 734-0000 734-0989 734-0986 734-0439 734-0440 734-0441 525-0122 734-0442 734-0443 734-0444 734-0445 734-0001
Round bottomed test tubes, Falcon®
Tubes Culture Tubes
For reliable containment of laboratory fluid samples.
  • Dual position snap-cap prevents sample loss with ease of handling, heavier gauge walls, and unique construction provide a secure, positive seal
  • Medical-style packaging materials
  • Tube dimensions and volumes are approximate
  • Widely used and referenced in laboratory protocols
  • Sterile, nonpyrogenicity tested to less than 0,1 EU/ml (Cat. No. 352008 nonsterile)
PS tubes: For procedures requiring high optical clarity