Imprimer
Tubes à essai
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
300500 300400 300700 300900 300705 300800 301700 300300 300100
212-3301EA 141 EUR
212-3301 212-3302 212-3303 212-3304 212-3305 212-3327 DELT301700 DELT300300 DELT300100
Tubes à essai
Tubes Tubes à essais
Clear polystyrene (PS).
  • Temperature resistance from −10 up to +70 °C (for short periods up to +80 °C)
  • Can be centrifuged up to 7500×g
  • Tubes 212-3304 graduated to 2,5/5,0 and 10 ml; tubes 212-3302 graduated to 1,25 and 5,0 ml
Precision moulded sample tubes for a wide variety of applications. With round base. Non sterile.