Imprimer
Transport and universal containers with caps, Nunc™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
333510 362820 364246 364882
391-8326EA 276 EUR
391-8326 391-8521 391-8101 391-8301
Transport and universal containers with caps, Nunc™
Récipients de collecte d'échantillon Conteneurs à usage universel
Transport and universal containers have passed the 95 kPa test described in UN packaging instructions 602/650 for transport of infectious substances and diagnostic specimens (except 391-8326). Universal containers are maximum RCF tested (RCF=Relative Centrifugal Force). Caps designed to avoid droplets adhering to the surface giving reduced risk of aerosols.
  • Safe handling and transport of blood, urine and faecal specimens
  • Inner and outer containers of high mechanical strength
  • Outer case available in transparent or opaque material
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904