Imprimer
PCR tubes, 0,2 ml, and cap strips, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PCR-0208-A PCR-0208-G PCR-0208-Y PCR-02CP-A PCR-02CP-C PCR-2CP-RT-C PCR-02-FCP-C PCR-12-FCP-C
731-4006EA 138 EUR
731-4006 731-4008 731-4012 731-4013 732-0680 732-0681 732-1375 731-0642
PCR tubes, 0,2 ml, and cap strips, Axygen®
Tubes Tubes PCR
Axygen® polypropylene 8/12-tube thin wall PCR tube strips and caps are specifically designed for PCR use and are ideal for thermal transfer applications. The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Made from highly polished moulds and free of lubricants, dyes, heavy metals, and fillers. Certified RNase, DNase, and pyrogen free to ensure the purity of your reaction.
  • Parois ultra-minces d'une épaisseur uniforme optimisant le transfert thermique
  • Langue à l'extrémité de chaque barrette pour une manipulation facilitée
  • Certifiés exempts de DNase, RNase et d'endotoxines
The polypropylene 8-strip tubes with individually attached caps come with added benefits that answer many customer needs. In addition to many of the features of a PCR tube strip, these feature a stronger connection between each tube and the caps attached. This minimises the risk of contamination and provides improved ease for customers when handling.