Imprimer
PCR tubes, 0,2 ml, with flat or domed cap, ABgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AB-0337 AB-0620 AB-0620/G AB-0620/P AB-0620/Y AB-0621 AB-0622 AB-0620/R AB-0620/B
732-4803EA 93.25 EUR
732-4803 732-4836 211-2262 732-0071 732-0072 732-0073 732-0074 211-2260 732-0070
PCR tubes, 0,2 ml, with flat or domed cap, ABgene®
Tubes PCR Tubes
PP.
  • 0,25 ml maximum tube capacity when closed
  • Integral snap-shut cap
  • Available with flat or domed caps
  • Certified free from DNase, RNase and Human DNA
Suitable for 0,2 ml thermal cycler blocks.