Imprimer
Microcentrifuge tubes, 1,5 ml, Secure-Lock™ cap
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T331-20B T331-20N T331-10N T331-10Y
720-1013EA 23.9 EUR
720-1013 720-1015 212-9613 212-9614
Microcentrifuge tubes, 1,5 ml, Secure-Lock™ cap
Tubes Microcentrifuge Tubes
PP, graduated at 0,5, 1,0 and at 1,5 ml, autoclavable with flat cap Secure-Lock™.
  • Can withstand centrifugation to 20000×g
  • Flat cap can be easily labelled and pierced
  • Certified RNase, DNase, pyrogen and DNA-free
  • With special locking clasp, Secure-Lock™ prevents tubes from opening during centrifugation, boiling, storing, freezing and shipping