Imprimer
Centrifuge tubes
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
525-0178EA 21.5 EUR
525-0178 525-0179 525-0180 525-0181 525-0182
Centrifuge tubes
Tubes Centrifuge Tubes
PTFE, round bottom, smooth surface, without screw cap.
  • Inert
  • Withstand forces up to 12 500×g
  • Very good chemical resistance
  • Suitable for use at high and low temperatures (–200 to +280 °C)
Suitable for use at high and low temperatures (–200 to +280 °C).