Imprimer
Temperature indicating strips
Fournisseur: TESTO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0646.1065 0646.1071 0646.1077 0646.1082 0646.1110 0646.1121
620-9151EA 34.3 EUR
620-9151 620-9152 620-9153 620-9154 620-9159 620-9161
Temperature indicating strips
Temperature Indicating Strips
These thermometer strips are made of self-adhesive foils with temperature sensitive elements for temperature control and regulation. They are used for measurements on moving and/or small parts and for long-term monitoring.
  • Irreversible change in colour within one second
  • Practical booklet with 50 thermometer strips
  • Storage at maximum 25 °C for up to two years; storage in a refrigerator is recommended
W×D: 15×14 mm