Imprimer
Microseringues, série 700
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
203073 203198 80300 80301 80308 80330 80336 80338 80350 80366 80383 80384 80391 80400 80401 80408 80430 80439 80500 80501 80508 80530 80600 80601 80630 80700 80701 80708 80730 80800 80801 80808 80830 80839 87900 87908 87919 87930 80608 7639-01 7634-01 7635-01 7636-01 7637-01 7638-01 7640-01 CAL80530 80660 87959 87994 80359 80550 80770 80551 CAL80383 CAL80300 CAL80600 CAL80400 CAL80500 CAL80730 CAL87930 CAL80700 CAL80366 80521 CAL87900 CAL80630 CAL80800 CAL80330 CAL80430 CAL87919 CAL80301
549-1317EA 124 EUR
549-1317 549-0571 549-1135 549-0221 613-1150 549-1137 549-0584 548-2291 549-1138 549-1140 549-0223 548-2292 549-1276 549-1144 549-0363 548-2293 549-1149 548-2294 549-1153 549-0225 548-2295 549-1154 549-1156 549-0227 549-1157 549-1159 549-0228 548-2304 549-1162 549-1166 549-0229 548-1963 549-1167 548-2307 549-1230 548-2328 549-0239 549-1231 548-2300 548-2276 548-2272 548-2273 548-2274 548-2275 548-1535 548-1536 HAMICAL80530 548-2303 549-0582 549-1306 549-0581 548-2298 548-2306 548-2299 548-2333 548-2332 548-2336 548-2334 548-2335 548-2337 HAMICAL87930 HAMICAL80700 548-1973 548-2296 HAMICAL87900 HAMICAL80630 HAMICAL80800 HAMICAL80330 HAMICAL80430 HAMICAL87919 HAMICAL80301
Microseringues, série 700
Seringues Seringues de chromatographie
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0,5 to 500 µl. The stainless steel plunger is manufactured to fit the glass barrel with a tolerance smaller than 100 millionths of an inch, resulting in unsurpassed syringe life. Each syringe is manufactured to achieve the highest level of accuracy and precision possible.
  • Fluid path is stainless steel and borosilicate glass
Cemented needle (N): Cemented needles for low volume syringes are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Removable needle (RN): The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. This termination also allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Luer tip (LT): Luer tip syringes feature needles that are removable and fit over a ground glass hub which is tapered in the shape of a male Luer. This termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination also increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
Needle point style 2: Sharp, beveled, curved, non-coring needle point recommended for septum penetration.
Needle point style 3: Blunt needle point for use with HPLC injection valves and for sample pipetting.
Needle point style 5: Conical needle with side port for penetration of septa, thin-gauged vinyls, and plastics without coring.
Needle point style AS: Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration.