Imprimer
Tapis de protection de surfaces, Labmat™
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F246750000 F246751000 F246750020
115-9101EA 0 EUR
115-9101 115-9102 115-2943
Tapis de protection de surfaces, Labmat™
Protection des surfaces Protections pour paillasse
Disposable Labmat™ bench liners speed clean-up and safeguard work surfaces from stains, spills and wear. Simply roll up and discard after contamination.
  • Highly absorbent upper surface traps up to 420 ml/m² of water, dyes, corrosive chemicals or radioactive solutions
  • Flexible polyethylene film backing is non-skid and prevents soak-through of most solutions
  • Available in 508 mm wide by 15,2 m rolls, or 508×610 mm pre-cut sheets
  • Rolls are easily cut while roll is retained in dispenser