Imprimer
Stoppers for standard ground joints
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
144405 144410 144415 144420 144425 144430 144435 144440 144445
217-9261EA 1.8 EUR
217-9261 217-9262 217-9263 217-9265 217-9266 217-9268 217-9270 217-9172 217-9174
Stoppers for standard ground joints
Stoppers PE/LDPE/HDPE Stoppers
PP or PE-LD, hollow, with sealing ribs and square or octagonal grip.
  • Good chemical resistance, PP stoppers are autoclavable (+121 °C)
  • Sealing rings for tight, leakproof fit
  • Square grip (up to sizes 29/32, PP) or octagonal grip (sizes 34/35 and larger, PE-LD) for easy release and removal of the stopper