Imprimer
Stoppers, red rubber
Fournisseur: LEY RUBBER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
RB005/S RB006/S RB007/S RB008/S RB009/S RB010/S RB011/S RB013/S RB015/S RB017/S RB019/S RB021/S RB023/S RB027/S RB029/S RB031/S RB033/S RB035/S RB067/S RB071/S RB075/S RB079/S RB086/S RB092/S RB099/S RB105/S RB037/S RB041/S RB043/S RB045/S RB053/S RB057/S RB062/S
LEYRRB005/SEA 48.3 EUR
LEYRRB005/S LEYRRB006/S LEYRRB007/S LEYRRB008/S LEYRRB009/S LEYRRB010/S LEYRRB011/S LEYRRB013/S LEYRRB015/S LEYRRB017/S LEYRRB019/S LEYRRB021/S LEYRRB023/S LEYRRB027/S LEYRRB029/S LEYRRB031/S LEYRRB033/S LEYRRB035/S LEYRRB067/S LEYRRB071/S LEYRRB075/S LEYRRB079/S LEYRRB086/S LEYRRB092/S LEYRRB099/S LEYRRB105/S LEYRRB037/S LEYRRB041/S LEYRRB043/S LEYRRB045/S LEYRRB053/S LEYRRB057/S LEYRRB062/S
Stoppers, red rubber
Bouchons Bouchons en caoutchouc
Solid, red rubber stoppers conforming to BS 2775. Highly elastic, for perfect sealing and adhesion.
  • Resistant to common laboratory chemicals
  • Provides a perfect seal and exceptional bonding
  • Solid form (pre-drilled holes available on request)
  • Resistant to temperatures from −5 to +70 °C (120 °C in humid atmosphere)