Imprimer
Stoppers, slightly conical, Versilic®
Fournisseur: Saint Gobain Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
261002 261034 261003
217-0877EA 358 EUR
217-0877 217-9475 217-0878
Stoppers, slightly conical, Versilic®
Stoppers Silicone Stoppers
These slightly conical translucent stoppers are made from silicone.
  • Biologically neutral, good chemical resistance
  • Odourless and tasteless
  • Resistant to ageing, ozone and repeated sterilisation
Temperature resistant from -60 to +200 °C (briefly up to 220 °C)